Phthalhydrazide, 99%
Phthalhydrazide, 99%
Phthalhydrazide, 99%
Thermo Scientific Chemicals

Phthalhydrazide, 99%

CAS: 1445-69-8 | C8H6N2O2 | 162.15 g/mol
数量:
25 g
100 g
パッケージ:
Plastic bottle
製品番号(カタログ番号) 131060250
価格(JPY)
-
数量:
25 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1445-69-8
IUPAC Name1,4,4a,8a-tetrahydrophthalazine-1,4-dione
Molecular FormulaC8H6N2O2
InChI KeyJNQUJPBFSQETPT-UHFFFAOYNA-N
SMILESO=C1N=NC(=O)C2C=CC=CC12
さらに表示
Appearance (Color)White
Infrared spectrumConforms
Titration with NaOH98.5 to 101.5 % (On dried substance)
Appearance (Form)Powder and/or chunks
Loss on drying=<1 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only