Piperazine hexahydrate, 98%
Piperazine hexahydrate, 98%
Piperazine hexahydrate, 98%
Thermo Scientific Chemicals

Piperazine hexahydrate, 98%

CAS: 142-63-2 | C4H10N2·6H2O | 194.23 g/mol
数量:
100 g
500 g
1 kg
5 kg
パッケージ:
すべて表示
Plastic bottle
製品番号(カタログ番号) 131300010
価格(JPY)
-
数量:
1 kg
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS142-63-2
IUPAC Namepiperazine hexahydrate
Molecular FormulaC4H22N2O6
InChI KeyAVRVZRUEXIEGMP-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.C1CNCCN1
さらに表示
Titration with HCl>=97.5 %
Appearance (Form)Adhering crystals
Infrared spectrumConforms
Appearance (Color)Transparent to white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only