2-Propanethiol, 98%
2-Propanethiol, 98%
2-Propanethiol, 98%
Thermo Scientific Chemicals

2-Propanethiol, 98%

CAS: 75-33-2 | C3H8S | 76.15 g/mol
数量:
50 mL
500 mL
2.5 L
パッケージ:
Glass bottle
製品番号(カタログ番号) 131440025
価格(JPY)
-
数量:
2.5 L
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS75-33-2
IUPAC Namepropane-2-thiol
Molecular FormulaC3H8S
InChI KeyKJRCEJOSASVSRA-UHFFFAOYSA-N
SMILESCC(C)S
さらに表示
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4240 to 1.4270 (20°C, 589 nm)
GC>=97.5 %
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only