n-Tridecane, 99+%
n-Tridecane, 99+%
n-Tridecane, 99+%
Thermo Scientific Chemicals

n-Tridecane, 99+%

CAS: 629-50-5 | C13H28 | 184.37 g/mol
数量:
25 mL
100 mL
500 mL
パッケージ:
Glass bottle
製品番号(カタログ番号) 139511000
価格(JPY)
-
数量:
100 mL
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS629-50-5
IUPAC Nametridecane
Molecular FormulaC13H28
InChI KeyIIYFAKIEWZDVMP-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCC
さらに表示
Appearance (Form)Liquid
GC>=99.0 %
Refractive index1.4240 to 1.4260 (20°C, 589 nm)
Appearance (Color)Clear colorless
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only