Triethylene glycol dimethyl ether, 99%, stabilized
Triethylene glycol dimethyl ether, 99%, stabilized
Triethylene glycol dimethyl ether, 99%, stabilized
Thermo Scientific Chemicals

Triethylene glycol dimethyl ether, 99%, stabilized

CAS: 112-49-2 | C8H18O4 | 178.23 g/mol
数量:
250 mL
1 L
2.5 L
パッケージ:
Glass bottle
製品番号(カタログ番号) 139600010
価格(JPY)
-
数量:
1 L
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS112-49-2
IUPAC Name2,5,8,11-tetraoxadodecane
Molecular FormulaC8H18O4
InChI KeyYFNKIDBQEZZDLK-UHFFFAOYSA-N
SMILESCOCCOCCOCCOC
さらに表示
Appearance (Form)Clear liquid
GC>=98.5 %
Water=<0.1 % (K.F.)
Acid value=<0.05 mg KOH/g
Infrared spectrumConforms
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only