2,3,3-Trimethylindolenine, 98%
2,3,3-Trimethylindolenine, 98%
2,3,3-Trimethylindolenine, 98%
Thermo Scientific Chemicals

2,3,3-Trimethylindolenine, 98%

CAS: 1640-39-7 | C11H13N | 159.23 g/mol
数量:
25 g
100 g
250 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 140161000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1640-39-7
IUPAC Name2,3,3-trimethyl-3H-indole
Molecular FormulaC11H13N
InChI KeyFLHJIAFUWHPJRT-UHFFFAOYSA-N
SMILESCC1=NC2=CC=CC=C2C1(C)C
さらに表示
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Liquid
Appearance (Color)Clear yellow to red-brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only