4-Methoxy-alpha-toluenethiol, 95%, Thermo Scientific Chemicals
4-Methoxy-alpha-toluenethiol, 95%, Thermo Scientific Chemicals
4-Methoxy-alpha-toluenethiol, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methoxy-alpha-toluenethiol, 95%, Thermo Scientific Chemicals

CAS: 6258-60-2 | Molecular Formula: C8H10OS | Molecular Weight: 154.23 g/mol
製品番号(カタログ番号)数量パッケージ
15095025025 gGlass bottle
製品番号(カタログ番号) 150950250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6258-60-2
IUPAC Name(4-methoxyphenyl)methanethiol
Molecular FormulaC8H10OS
InChI KeyPTDVPWWJRCOIIO-UHFFFAOYSA-N
SMILESCOC1=CC=C(CS)C=C1
さらに表示
Appearance (Color)Clear colorless to light yellow
Refractive index1.5720 to 1.5780 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=94 %

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only