4-Nitrobenzenesulfonamide, 97%
4-Nitrobenzenesulfonamide, 97%
4-Nitrobenzenesulfonamide, 97%
Thermo Scientific Chemicals

4-Nitrobenzenesulfonamide, 97%

CAS: 6325-93-5 | C6H6N2O4S | 202.18 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 153020050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6325-93-5
IUPAC Name4-nitrobenzene-1-sulfonamide
Molecular FormulaC6H6N2O4S
InChI KeyQWKKYJLAUWFPDB-UHFFFAOYSA-N
SMILESNS(=O)(=O)C1=CC=C(C=C1)[N+]([O-])=O
さらに表示
Infrared spectrumConforms
Melting point175°C to 181°C
Appearance (Form)Crystalline powder or crystals
HPLC>=96.0 %
Appearance (Color)White to beige to tan
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only