2-Chlorobenzaldehyde, 99%
2-Chlorobenzaldehyde, 99%
2-Chlorobenzaldehyde, 99%
Thermo Scientific Chemicals

2-Chlorobenzaldehyde, 99%

CAS: 89-98-5 | C7H5ClO | 140.57 g/mol
数量:
100 mL
250 mL
500 mL
1 L
パッケージ:
Glass bottle
製品番号(カタログ番号) 154615000
価格(JPY)
-
数量:
500 mL
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS89-98-5
IUPAC Name2-chlorobenzaldehyde
Molecular FormulaC7H5ClO
InChI KeyFPYUJUBAXZAQNL-UHFFFAOYSA-N
SMILESClC1=CC=CC=C1C=O
さらに表示
Appearance (Color)Clear colorless to light yellow
Free acid=<1 % (as chlorobenzoic acid)
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only