2,3-Dihydroxybenzoic acid, 99%
2,3-Dihydroxybenzoic acid, 99%
2,3-Dihydroxybenzoic acid, 99%
Thermo Scientific Chemicals

2,3-Dihydroxybenzoic acid, 99%

CAS: 303-38-8 | C7H6O4 | 154.12 g/mol
製品番号(カタログ番号)数量パッケージ
15517025025 gGlass bottle
製品番号(カタログ番号) 155170250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS303-38-8
IUPAC Name2,3-dihydroxybenzoic acid
Molecular FormulaC7H6O4
InChI KeyGLDQAMYCGOIJDV-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC(O)=C1O
さらに表示
Appearance (Color)Beige to pink to grey to brown
Melting point204°C to 208°C
HPLC>=98.5 %
Infrared spectrumConforms
Titration with NaOH>=98.5 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only