N,N,N',N'-Tetraethylethylenediamine, 99+%
N,N,N',N'-Tetraethylethylenediamine, 99+%
N,N,N',N'-Tetraethylethylenediamine, 99+%
Thermo Scientific Chemicals

N,N,N',N'-Tetraethylethylenediamine, 99+%

CAS: 150-77-6 | C10H26N2 | 174.33 g/mol
数量:
10 mL
100 mL
500 mL
製品番号(カタログ番号) 155590100
価格(JPY)
-
数量:
10 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS150-77-6
IUPAC Name[2-(diethylazaniumyl)ethyl]diethylazanium
Molecular FormulaC10H26N2
InChI KeyDIHKMUNUGQVFES-UHFFFAOYSA-P
SMILESCC[NH+](CC)CC[NH+](CC)CC
さらに表示
Appearance (Form)Liquid
GC>=99.0 %
Refractive index1.4333 to 1.4353 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only