Methylcyclopentadiene dimer, 90%, stabilized
Methylcyclopentadiene dimer, 90%, stabilized
Methylcyclopentadiene dimer, 90%, stabilized
Thermo Scientific Chemicals

Methylcyclopentadiene dimer, 90%, stabilized

CAS: 26472-00-4 | C12H16 | 160.26 g/mol
数量:
250 g
1 kg
2.5 kg
パッケージ:
Glass bottle
製品番号(カタログ番号) 156852500
価格(JPY)
-
数量:
250 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS26472-00-4
IUPAC Name4,9-dimethyltricyclo[5.2.1.0²,⁶]deca-3,8-diene
Molecular FormulaC12H16
InChI KeyIYQYZZHQSZMZIG-UHFFFAOYNA-N
SMILESCC1=CC2C(C1)C1CC2C(C)=C1
さらに表示
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4940 to 1.5020 (20°C, 589 nm)
GC>=88 % (Sum of isomers)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only