2-Furoic acid hydrazide, 98%
2-Furoic acid hydrazide, 98%
2-Furoic acid hydrazide, 98%
Thermo Scientific Chemicals

2-Furoic acid hydrazide, 98%

CAS: 3326-71-4 | C5H6N2O2 | 126.115 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 157160250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS3326-71-4
IUPAC Namefuran-2-carbohydrazide
Molecular FormulaC5H6N2O2
InChI KeySKTSVWWOAIAIKI-UHFFFAOYSA-N
SMILESNNC(=O)C1=CC=CO1
さらに表示
Appearance (Form)Crystals
Melting point76°C to 80°C
Appearance (Color)Beige
GC>=97.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only