DL-2-Methyl-1-butanol, 98%
DL-2-Methyl-1-butanol, 98%
DL-2-Methyl-1-butanol, 98%
Thermo Scientific Chemicals

DL-2-Methyl-1-butanol, 98%

CAS: 137-32-6 | C5H12O | 88.15 g/mol
数量:
100 mL
1 L
2.5 L
パッケージ:
Glass bottle
製品番号(カタログ番号) 158271000
価格(JPY)
-
数量:
100 mL
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS137-32-6
IUPAC Name2-methylbutan-1-ol
Molecular FormulaC5H12O
InChI KeyQPRQEDXDYOZYLA-UHFFFAOYNA-N
SMILESCCC(C)CO
さらに表示
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4090 to 1.4120 (20°C, 589 nm)
GC>=97.5 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only