1-Adamantanamine, 96%
1-Adamantanamine, 96%
1-Adamantanamine, 96%
Thermo Scientific Chemicals

1-Adamantanamine, 96%

CAS: 768-94-5 | C10H17N | 151.25 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 160470250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS768-94-5
IUPAC Nameadamantan-1-amine
Molecular FormulaC10H17N
InChI KeyDKNWSYNQZKUICI-UHFFFAOYSA-N
SMILESNC12CC3CC(CC(C3)C1)C2
さらに表示
Titration with HClO495.0 to 105.0 % (On dry substance)
Appearance (Color)White to cream
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Water=<15 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Adamantanamine acts as a catalyst in enantioselective strecker reaction of phosphinoyl imines. It is also useful for making adamantane derivatives, i.e. amantadine hydrochloride. It is used in the pharmaceutical industry as antiviral and an antiparkinsonian drug.

Solubility
Soluble in organic solvents. Insoluble in water.

Notes
Use only in a chemical fume hood. Keep away from sources of ignition. Incompatible with oxidizing agent.
RUO – Research Use Only