trans-Ethyl crotonate, 96%
trans-Ethyl crotonate, 96%
trans-Ethyl crotonate, 96%
Thermo Scientific Chemicals

trans-Ethyl crotonate, 96%

CAS: 623-70-1 | C6H10O2 | 114.14 g/mol
数量:
100 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 161381000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS623-70-1
IUPAC Nameethyl (2E)-but-2-enoate
Molecular FormulaC6H10O2
InChI KeyZFDIRQKJPRINOQ-HWKANZROSA-N
SMILESCCOC(=O)\C=C\C
さらに表示
Infrared spectrumConforms
Refractive index1.4220 to 1.4280 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear colorless
GC>=95 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only