5-Bromosalicylic acid, 98%
5-Bromosalicylic acid, 98%
5-Bromosalicylic acid, 98%
Thermo Scientific Chemicals

5-Bromosalicylic acid, 98%

CAS: 89-55-4 | C7H5BrO3 | 217.02 g/mol
数量:
50 g
250 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 162140500
価格(JPY)
-
数量:
50 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS89-55-4
IUPAC Name5-bromo-2-hydroxybenzoic acid
Molecular FormulaC7H5BrO3
InChI KeyIEJOONSLOGAXNO-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(Br)=CC=C1O
さらに表示
Infrared spectrumConforms
Appearance (Color)White to light beige
Melting point164°C to 172°C
Titration with NaOH97.5 to 102.5 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only