5-Methylsalicylic acid, 98%
5-Methylsalicylic acid, 98%
5-Methylsalicylic acid, 98%
Thermo Scientific Chemicals

5-Methylsalicylic acid, 98%

CAS: 89-56-5 | C8H8O3 | 152.15 g/mol
製品番号(カタログ番号)数量パッケージ
16386050050 gGlass bottle
製品番号(カタログ番号) 163860500
価格(JPY)
-
数量:
50 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS89-56-5
IUPAC Name2-hydroxy-5-methylbenzoic acid
Molecular FormulaC8H8O3
InChI KeyDLGBEGBHXSAQOC-UHFFFAOYSA-N
SMILESCC1=CC=C(O)C(=C1)C(O)=O
さらに表示
Titration with NaOH>=97.5 %
Melting point151°C to 155°C
Appearance (Color)White to beige
Infrared spectrumConforms
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only