Thermo Scientific Chemicals

3',4'-Dimethoxyacetophenone, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): 168851000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

3',4'-Dimethoxyacetophenone, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): 168851000
100 g, Glass bottle, Each
数量
パッケージ
製品番号(カタログ番号): 168851000

化学物質識別子

CAS
1131-62-0
IUPAC Name
1-(3,4-dimethoxyphenyl)ethan-1-one
Molecular Formula
C10H12O3
InChI Key
IQZLUWLMQNGTIW-UHFFFAOYSA-N
SMILES
COC1=CC=C(C=C1OC)C(C)=O
Appearance (Color)
Yellow to beige
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
GC
>=97.5 %
Melting point
48°C to 52°C

概要

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text