Triethanolamine hydrochloride, +99%, Thermo Scientific Chemicals
Triethanolamine hydrochloride, +99%, Thermo Scientific Chemicals
Triethanolamine hydrochloride, +99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triethanolamine hydrochloride, +99%, Thermo Scientific Chemicals

数量:
100 g
500 g
2.5 kg
パッケージ:
すべて表示
Plastic bottle
製品番号(カタログ番号) 170050025
価格(JPY)
-
数量:
2.5 kg
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS637-39-8
IUPAC Namehydrogen 2-[bis(2-hydroxyethyl)amino]ethan-1-ol chloride
Molecular FormulaC6H16ClNO3
InChI KeyHHLJUSLZGFYWKW-UHFFFAOYSA-N
SMILES[H+].[Cl-].OCCN(CCO)CCO
さらに表示
Titration Argentometric>=99.0 %
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Loss on drying=<1.0 % (105°C)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only