3-Aminopyrazole, 98%
3-Aminopyrazole, 98%
3-Aminopyrazole, 98%
Thermo Scientific Chemicals

3-Aminopyrazole, 98%

CAS: 1820-80-0 | C3H5N3 | 83.09 g/mol
数量:
2.5 g
10 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 171030025
価格(JPY)
-
数量:
2.5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1820-80-0
IUPAC Name1H-pyrazol-5-amine
Molecular FormulaC3H5N3
InChI KeyJVVRJMXHNUAPHW-UHFFFAOYSA-N
SMILESNC1=CC=NN1
さらに表示
Appearance (Color)Colorless to light brown
Appearance (Form)Liquid or fused solid
Infrared spectrumConforms
Titration with HClO497.5 to 102.5 %
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only