2-Fluorobenzylamine, 96%
2-Fluorobenzylamine, 96%
2-Fluorobenzylamine, 96%
Thermo Scientific Chemicals

2-Fluorobenzylamine, 96%

CAS: 89-99-6 | C7H8FN | 125.15 g/mol
数量:
1 g
10 g
50 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 171940010
価格(JPY)
-
数量:
1 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS89-99-6
IUPAC Name1-(2-fluorophenyl)methanamine
Molecular FormulaC7H8FN
InChI KeyLRFWYBZWRQWZIM-UHFFFAOYSA-N
SMILESNCC1=CC=CC=C1F
さらに表示
Appearance (Color)Clear colorless to orange
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5150 to 1.5190 (20°C, 589 nm)
GC>=95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only