1,3-Dichloroacetone, 96%
1,3-Dichloroacetone, 96%
1,3-Dichloroacetone, 96%
Thermo Scientific Chemicals

1,3-Dichloroacetone, 96%

CAS: 534-07-6 | C3H4Cl2O | 126.96 g/mol
数量:
25 g
50 g
250 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 173630500
価格(JPY)
-
数量:
50 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS534-07-6
IUPAC Name1,3-dichloropropan-2-one
Molecular FormulaC3H4Cl2O
InChI KeySUNMBRGCANLOEG-UHFFFAOYSA-N
SMILESClCC(=O)CCl
さらに表示
Appearance (Color)White to light brown
Infrared spectrumConforms
Melting point38°C to 44°C
Appearance (Form)Crystalline powder or crystals and/or chunks
GC>=95 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only