4-Hydroxy-2,2,6,6-tetramethyl-piperidinooxy, 98%, free radical
4-Hydroxy-2,2,6,6-tetramethyl-piperidinooxy, 98%, free radical
4-Hydroxy-2,2,6,6-tetramethyl-piperidinooxy, 98%, free radical
Thermo Scientific Chemicals

4-Hydroxy-2,2,6,6-tetramethyl-piperidinooxy, 98%, free radical

CAS: 2226-96-2 | C9H18NO2 | 172.25 g/mol
数量:
1 g
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 175050050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2226-96-2
IUPAC Name(4-hydroxy-2,2,6,6-tetramethylpiperidin-1-yl)oxidanyl
Molecular FormulaC9H18NO2
InChI KeyUZFMOKQJFYMBGY-UHFFFAOYSA-N
SMILESCC1(C)CC(O)CC(C)(C)N1[O]
さらに表示
Appearance (Color)Orange
Appearance (Form)Crystalline powder or crystals or flakes
Infrared spectrumConforms
GC>=97.5 %
Melting point69°C to 73°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Polymerization reagent
RUO – Research Use Only