3-Quinolinecarboxylic acid, 98%
3-Quinolinecarboxylic acid, 98%
3-Quinolinecarboxylic acid, 98%
Thermo Scientific Chemicals

3-Quinolinecarboxylic acid, 98%

CAS: 6480-68-8 | C10H7NO2 | 173.17 g/mol
数量:
1 g
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 175700050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6480-68-8
IUPAC Namequinoline-3-carboxylic acid
Molecular FormulaC10H7NO2
InChI KeyDJXNJVFEFSWHLY-UHFFFAOYSA-N
SMILESOC(=O)C1=CN=C2C=CC=CC2=C1
さらに表示
Appearance (Color)White to light beige
Infrared spectrumConforms
Titration with HClO4>=97.5 %
Appearance (Form)Powder
Melting point277°C to 283°C
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only