Starch, for analysis, soluble
Starch, for analysis, soluble
Starch, for analysis, soluble
Thermo Scientific Chemicals

Starch, for analysis, soluble

CAS: 9005-84-9 | C12H22O11 | 342.297 g/mol
数量:
250 g
1 kg
5 kg
パッケージ:
すべて表示
Plastic bottle
製品番号(カタログ番号) 177130010
価格(JPY)
-
数量:
1 kg
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS9005-84-9
IUPAC Name(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
Molecular FormulaC12H22O11
InChI KeyGUBGYTABKSRVRQ-ASMJPISFSA-N
SMILESOC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O
さらに表示
pH5 to 7.5 (2 % at 25°C)
SensitivityPasses test
IdentificationPositive
Appearance (Color)White
Appearance (Form)Powder
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only