2-Mercaptopyridine-N-oxide, 99%
2-Mercaptopyridine-N-oxide, 99%
2-Mercaptopyridine-N-oxide, 99%
Thermo Scientific Chemicals

2-Mercaptopyridine-N-oxide, 99%

CAS: 1121-31-9 | C5H5NOS | 127.16 g/mol
数量:
1 g
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 182670010
価格(JPY)
-
数量:
1 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1121-31-9
IUPAC Name1-hydroxy-1,2-dihydropyridine-2-thione
Molecular FormulaC5H5NOS
InChI KeyYBBJKCMMCRQZMA-UHFFFAOYSA-N
SMILESON1C=CC=CC1=S
さらに表示
Appearance (Color)Off-white to yellow to yellow-green to beige to
Infrared spectrumConforms
Appearance (Form)Powder or crystals or chunks
Titration Iodometric>=98.5 %
Appearance (Color)grey-green
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only