3-Phenoxybenzyl alcohol, 98%
3-Phenoxybenzyl alcohol, 98%
3-Phenoxybenzyl alcohol, 98%
Thermo Scientific Chemicals

3-Phenoxybenzyl alcohol, 98%

CAS: 13826-35-2 | C13H12O2 | 200.24 g/mol
数量:
5 g
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 183770050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS13826-35-2
IUPAC Name(3-phenoxyphenyl)methanol
Molecular FormulaC13H12O2
InChI KeyKGANAERDZBAECK-UHFFFAOYSA-N
SMILESOCC1=CC=CC(OC2=CC=CC=C2)=C1
さらに表示
Appearance (Color)Clear colorless to yellow
GC>=97.5 %
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.5910 to 1.5950 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only