Ethyl nitroacetate, 98%
Ethyl nitroacetate, 98%
Ethyl nitroacetate, 98%
Thermo Scientific Chemicals

Ethyl nitroacetate, 98%

CAS: 626-35-7 | C4H7NO4 | 133.1 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) 185171000
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS626-35-7
IUPAC Nameethyl 2-nitroacetate
Molecular FormulaC4H7NO4
InChI KeyFTKASJMIPSSXBP-UHFFFAOYSA-N
SMILESCCOC(=O)C[N+]([O-])=O
さらに表示
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
Refractive index1.4235 to 1.4255 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only