Fast Blue BB, 98%, pure
Fast Blue BB, 98%, pure
Fast Blue BB, 98%, pure
Thermo Scientific Chemicals

Fast Blue BB, 98%, pure

CAS: 120-00-3 | C17H20N2O3 | 300.358 g/mol
数量:
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 191571000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS120-00-3
IUPAC NameN-(4-amino-2,5-diethoxyphenyl)benzamide
Molecular FormulaC17H20N2O3
InChI KeyCNXZLZNEIYFZGU-UHFFFAOYSA-N
SMILESCCOC1=CC(NC(=O)C2=CC=CC=C2)=C(OCC)C=C1N
さらに表示
Infrared spectrumConforms
Appearance (Color)Grey to brown
HPLC>=97.5 %
Appearance (Form)Powder and/or chunks
UV spectrumComplies
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only