Kojic acid, 99%
Kojic acid, 99%
Kojic acid, 99%
Thermo Scientific Chemicals

Kojic acid, 99%

CAS: 501-30-4 | C6H6O4 | 142.11 g/mol
数量:
5 g
25 g
100 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 205225000
価格(JPY)
-
数量:
500 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS501-30-4
IUPAC Name5-hydroxy-2-(hydroxymethyl)-4H-pyran-4-one
Molecular FormulaC6H6O4
InChI KeyBEJNERDRQOWKJM-UHFFFAOYSA-N
SMILESOCC1=CC(=O)C(O)=CO1
さらに表示
Appearance (Form)Crystalline powder
Melting point151°C to 156°C
Infrared spectrumConforms
Appearance (Color)White to beige to light brown
Titration with NaOH>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only