Phosphomolybdic acid hydrate, 80%
Phosphomolybdic acid hydrate, 80%
Phosphomolybdic acid hydrate, 80%
Thermo Scientific Chemicals

Phosphomolybdic acid hydrate, 80%

CAS: 51429-74-4 | H3Mo12O40P | 1825.40 g/mol
数量:
25 g
100 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 206380250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS51429-74-4
IUPAC Name12-Molybdophosphoric acid
Molecular FormulaH3Mo12O40P
Molecular Weight (g/mol)1825.40
Synonymphosphoric acid dodecamolybdenum hexatriacontaoxidandiide
さらに表示
Chloride (Cl)=<200 ppm
Iron (Fe)=<50 ppm
Sulfate (SO4)=<250 ppm
Appearance (Color)Yellow to yellow-orange
Assay>=47.5 % (Mo)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only