Potassium bromide, 99+%, for spectroscopy, IR grade
Potassium bromide, 99+%, for spectroscopy, IR grade
Potassium bromide, 99+%, for spectroscopy, IR grade
Thermo Scientific Chemicals

Potassium bromide, 99+%, for spectroscopy, IR grade

CAS: 7758-02-3 | BrK | 119.00 g/mol
数量:
25 g, 25 g
100 g
500 g
パッケージ:
すべて表示
Plastic bottle, Plastic bottle
製品番号(カタログ番号) 206390250
価格(JPY)
-
数量:
25 g, 25 g
パッケージ:
Plastic bottle, Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7758-02-3
IUPAC Namepotassium bromide
Molecular FormulaBrK
InChI KeyIOLCXVTUBQKXJR-UHFFFAOYSA-M
SMILES[K+].[Br-]
さらに表示
Application testPasses test
Application testPasses test
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For IR spectroscopy
RUO – Research Use Only