2-Methylbenzofuran, 96%
2-Methylbenzofuran, 96%
2-Methylbenzofuran, 96%
Thermo Scientific Chemicals

2-Methylbenzofuran, 96%

CAS: 4265-25-2 | C9H8O | 132.16 g/mol
数量:
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 208400050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4265-25-2
IUPAC Name2-methyl-1-benzofuran
Molecular FormulaC9H8O
InChI KeyGBGPVUAOTCNZPT-UHFFFAOYSA-N
SMILESCC1=CC2=CC=CC=C2O1
さらに表示
Appearance (Color)Colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=95.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only