Citric acid, monosodium salt, 99%, pure, anhydrous
Citric acid, monosodium salt, 99%, pure, anhydrous
Citric acid, monosodium salt, 99%, pure, anhydrous
Thermo Scientific Chemicals

Citric acid, monosodium salt, 99%, pure, anhydrous

CAS: 18996-35-5 | C6H7NaO7 | 214.11 g/mol
数量:
100 g
500 g
2.5 kg
パッケージ:
すべて表示
Glass bottle
製品番号(カタログ番号) 215331000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS18996-35-5
IUPAC Namesodium 3-carboxy-2-(carboxymethyl)-2-hydroxypropanoate
Molecular FormulaC6H7NaO7
InChI KeyHWPKGOGLCKPRLZ-UHFFFAOYSA-M
SMILES[Na+].OC(=O)CC(O)(CC(O)=O)C([O-])=O
さらに表示
Titration with HClO4>=98.5 %
Appearance (Form)Crystalline powder
Appearance (Color)White
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only