4-Acetophenetidide, 97%
4-Acetophenetidide, 97%
4-Acetophenetidide, 97%
Thermo Scientific Chemicals

4-Acetophenetidide, 97%

CAS: 62-44-2 | C10H13NO2 | 179.22 g/mol
数量:
100 g
500 g
2.5 kg
製品番号(カタログ番号) 216591000
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS62-44-2
IUPAC NameN-(4-ethoxyphenyl)acetamide
Molecular FormulaC10H13NO2
InChI KeyCPJSUEIXXCENMM-UHFFFAOYSA-N
SMILESCCOC1=CC=C(NC(C)=O)C=C1
さらに表示
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=96.0 %
Melting point133°C to 138°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only