Methyl sulfoxide-d6, for NMR, packaged in 0.50 ml ampoules, 100.0 atom % D
Methyl sulfoxide-d6, for NMR, packaged in 0.50 ml ampoules, 100.0 atom % D
Methyl sulfoxide-d6, for NMR, packaged in 0.50 ml ampoules, 100.0 atom % D
Thermo Scientific Chemicals

Methyl sulfoxide-d6, for NMR, packaged in 0.50 ml ampoules, 100.0 atom % D

CAS: 2206-27-1 | C2H6OS | 84.17 g/mol
製品番号(カタログ番号)数量
2173400505 mL
製品番号(カタログ番号) 217340050
価格(JPY)
-
数量:
5 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2206-27-1
IUPAC Name(²H₃)methanesulfinyl(²H₃)methane
Molecular FormulaC2H6OS
InChI KeyIAZDPXIOMUYVGZ-WFGJKAKNSA-N
SMILES[2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
さらに表示
Appearance (Form)Liquid
Water=<0.02 % (Coulometric)
D-Enrichment>=99.95 %
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only