Dimethyl maleate, 96%
Dimethyl maleate, 96%
Dimethyl maleate, 96%
Thermo Scientific Chemicals

Dimethyl maleate, 96%

CAS: 624-48-6 | C6H8O4 | 144.13 g/mol
数量:
250 mL
1 L
5 L
パッケージ:
すべて表示
Plastic drum
製品番号(カタログ番号) 218460051
価格(JPY)
-
数量:
5 L
パッケージ:
Plastic drum
一括またはカスタム形式をリクエストする
化学物質識別子
CAS624-48-6
IUPAC Name1,4-dimethyl (2Z)-but-2-enedioate
Molecular FormulaC6H8O4
InChI KeyLDCRTTXIJACKKU-ARJAWSKDSA-N
SMILESCOC(=O)\C=C/C(=O)OC
さらに表示
Color scale=<20 APHA
Appearance (Form)Clear liquid
Infrared spectrumConforms
Water=<0.2 % (K.F.)
GC>=95 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only