1-Butanethiol, 97%
1-Butanethiol, 97%
1-Butanethiol, 97%
Thermo Scientific Chemicals

1-Butanethiol, 97%

CAS: 109-79-5 | C4H10S | 90.18 g/mol
数量:
50 mL
250 mL
1 L
パッケージ:
Glass bottle
製品番号(カタログ番号) 220622500
価格(JPY)
-
数量:
250 mL
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS109-79-5
IUPAC Namebutane-1-thiol
Molecular FormulaC4H10S
InChI KeyWQAQPCDUOCURKW-UHFFFAOYSA-N
SMILESCCCCS
さらに表示
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.4420 to 1.4440 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only