Tetraethylammonium tetrafluoroborate, 99%
Tetraethylammonium tetrafluoroborate, 99%
Tetraethylammonium tetrafluoroborate, 99%
Thermo Scientific Chemicals

Tetraethylammonium tetrafluoroborate, 99%

CAS: 429-06-1 | C8H20BF4N | 217.06 g/mol
数量:
25 g
100 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 221611000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS429-06-1
IUPAC Nametetraethylazanium; tetrafluoroboranuide
Molecular FormulaC8H20BF4N
InChI KeyXJRAKUDXACGCHA-UHFFFAOYSA-N
SMILESF[B-](F)(F)F.CC[N+](CC)(CC)CC
さらに表示
Appearance (Form)Powder or crystals and/or lumps
Elemental Analysis>=98.5 % (ex C)
Appearance (Color)White
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only