2-Bromothiophenol, 97%
2-Bromothiophenol, 97%
2-Bromothiophenol, 97%
Thermo Scientific Chemicals

2-Bromothiophenol, 97%

CAS: 6320-02-1 | C6H5BrS | 189.07 g/mol
数量:
1 g
10 g
50 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 221980010
価格(JPY)
-
数量:
1 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6320-02-1
IUPAC Name2-bromobenzene-1-thiol
Molecular FormulaC6H5BrS
InChI KeyYUQUNWNSQDULTI-UHFFFAOYSA-N
SMILESSC1=CC=CC=C1Br
さらに表示
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Refractive index1.6340 to 1.6370 (20°C, 589 nm)
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only