3-Iodoanisole, 97%
3-Iodoanisole, 97%
3-Iodoanisole, 97%
Thermo Scientific Chemicals

3-Iodoanisole, 97%

CAS: 766-85-8 | C7H7IO | 234.04 g/mol
数量:
5 g
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 222021000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS766-85-8
IUPAC Name1-iodo-3-methoxybenzene
Molecular FormulaC7H7IO
InChI KeyRSHBAGGASAJQCH-UHFFFAOYSA-N
SMILESCOC1=CC=CC(I)=C1
さらに表示
Appearance (Color)Clear yellow
Appearance (Form)Liquid
AppearanceMay darken on storage
Infrared spectrumConforms
GC>=96.0 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only