Diphenylamine-4-sulfonic acid, sodium salt, pure, redox-indicator grade
Diphenylamine-4-sulfonic acid, sodium salt, pure, redox-indicator grade
Diphenylamine-4-sulfonic acid, sodium salt, pure, redox-indicator grade
Thermo Scientific Chemicals

Diphenylamine-4-sulfonic acid, sodium salt, pure, redox-indicator grade

CAS: 6152-67-6 | C12H11NNaO3S | 272.27 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 222220050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6152-67-6
IUPAC Name4-(phenylamino)benzene-1-sulfonic acid sodium
Molecular FormulaC12H11NNaO3S
InChI KeyVMVSDVIYIMNZBS-UHFFFAOYSA-N
SMILES[Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1
さらに表示
Appearance (Color)Off-white to beige to light grey
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Sensitivity(as indicator) Passes test
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only