1-Bromo-2-butanone, 90%, stabilized, Thermo Scientific Chemicals
1-Bromo-2-butanone, 90%, stabilized, Thermo Scientific Chemicals
1-Bromo-2-butanone, 90%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Bromo-2-butanone, 90%, stabilized, Thermo Scientific Chemicals

数量:
1 g
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 222450050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS816-40-0
IUPAC Name1-bromobutan-2-one
Molecular FormulaC4H7BrO
InChI KeyCCXQVBSQUQCEEO-UHFFFAOYSA-N
SMILESCCC(=O)CBr
さらに表示
Appearance (Color)Colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=88 %
Refractive index1.4650 to 1.4690 (20°C, 589 nm)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only