2-Pentanone, 99+%, purified by redistillation, AcroSeal™
2-Pentanone, 99+%, purified by redistillation, AcroSeal™
2-Pentanone, 99+%, purified by redistillation, AcroSeal™
Thermo Scientific Chemicals

2-Pentanone, 99+%, purified by redistillation, AcroSeal™

CAS: 107-87-9 | C5H10O | 86.13 g/mol
数量:
100 mL
1 L
パッケージ:
AcroSeal™ Glass bottle
製品番号(カタログ番号) 224130010
価格(JPY)
-
数量:
1 L
パッケージ:
AcroSeal™ Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS107-87-9
IUPAC Namepentan-2-one
Molecular FormulaC5H10O
InChI KeyXNLICIUVMPYHGG-UHFFFAOYSA-N
SMILESCCCC(C)=O
さらに表示
Appearance (Form)Liquid
Appearance (Color)Clear colorless
GC>=99.0 %
Infrared spectrumConforms
Color scale=<10 APHA
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only