6-(Methylthio)purine, 98%
6-(Methylthio)purine, 98%
6-(Methylthio)purine, 98%
Thermo Scientific Chemicals

6-(Methylthio)purine, 98%

CAS: 50-66-8 | C6H6N4S | 166.20 g/mol
製品番号(カタログ番号)数量パッケージ
2265600101 gGlass bottle
製品番号(カタログ番号) 226560010
価格(JPY)
-
数量:
1 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS50-66-8
IUPAC Name6-(methylsulfanyl)-7H-purine
Molecular FormulaC6H6N4S
InChI KeyUIJIQXGRFSPYQW-UHFFFAOYSA-N
SMILESCSC1=C2NC=NC2=NC=N1
さらに表示
Infrared spectrumConforms
Titration with NaOH>=97.5 %
Melting point219°C to 223°C
Appearance (Form)Powder
Appearance (Color)White to light yellow to green-beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only