L-Methionine sulfoximine, 98+%
L-Methionine sulfoximine, 98+%
L-Methionine sulfoximine, 98+%
Thermo Scientific Chemicals

L-Methionine sulfoximine, 98+%

CAS: 15985-39-4 | C5H12N2O3S | 180.22 g/mol
数量:
250 mg
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 227200010
価格(JPY)
-
数量:
1 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS15985-39-4
IUPAC Name(2S)-2-azaniumyl-4-[imino(methyl)oxo-λ⁶-sulfanyl]butanoate
Molecular FormulaC5H12N2O3S
InChI KeySXTAYKAGBXMACB-DPVSGNNYSA-N
SMILESCS(=N)(=O)CC[C@H]([NH3+])C([O-])=O
さらに表示
Appearance (Form)Crystalline powder
Titration with HClO498.0 to 102.0 %
Appearance (Color)White to almost white
Specific optical rotation+12° to +15° (20°C, 589 nm) (c=1, H2O)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only