Erioglaucine disodium salt, pure
Erioglaucine disodium salt, pure
Erioglaucine disodium salt, pure
Thermo Scientific Chemicals

Erioglaucine disodium salt, pure

CAS: 3844-45-9 | C37H34N2Na2O9S3 | 792.84 g/mol
製品番号(カタログ番号)数量パッケージ
2297300505 gGlass bottle
製品番号(カタログ番号) 229730050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS3844-45-9
IUPAC Namedisodium bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(2-sulfonatophenyl)methylium
Molecular FormulaC37H34N2Na2O9S3
InChI KeySGHZXLIDFTYFHQ-UHFFFAOYSA-L
SMILES[Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=CC=C1S([O-])(=O)=O
さらに表示
Appearance (Form)Crystalline powder
Loss on drying=<10 % (105°C)
UV Lambda max625 to 630 nm
Absorption Ratio P-15/P+151.10 to 1.25
Specific absorbance A (1%/1cm)>=1000 (at lambda max.)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only