3-Hydroxypicolinic acid, 98%
3-Hydroxypicolinic acid, 98%
3-Hydroxypicolinic acid, 98%
Thermo Scientific Chemicals

3-Hydroxypicolinic acid, 98%

CAS: 874-24-8 | C6H4NO3 | 138.10 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 241050050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS874-24-8
IUPAC Name3-hydroxypyridine-2-carboxylate
Molecular FormulaC6H4NO3
InChI KeyBRARRAHGNDUELT-UHFFFAOYSA-M
SMILESOC1=CC=CN=C1C([O-])=O
さらに表示
HPLC>=97.5 %
Appearance (Form)Powder
Melting point208°C to 214°C
Appearance (Color)Beige to brown
Infrared spectrumConforms
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only