Benzo-18-crown-6, 97%
Benzo-18-crown-6, 97%
Benzo-18-crown-6, 97%
Thermo Scientific Chemicals

Benzo-18-crown-6, 97%

CAS: 14098-24-9 | C16H24O6 | 312.34 g/mol
数量:
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 243980010
価格(JPY)
-
数量:
1 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS14098-24-9
IUPAC Name2,3,5,6,8,9,11,12,14,15-decahydro-1,4,7,10,13,16-benzohexaoxacyclooctadecine
Molecular FormulaC16H24O6
InChI KeyDSFHXKRFDFROER-UHFFFAOYSA-N
SMILESC1COCCOCCOC2=CC=CC=C2OCCOCCO1
さらに表示
Melting point40°C to 45°C
Appearance (Color)White to off-white
Appearance (Form)Powder or crystals and/or chunks
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only